Learn More
Horst™ HBQ Model Very Flexible Heating Tape
Brand:  Horst™ 20403
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 10322462
  • 450.50€ / Each
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
Récupel-bijdrage uitgesloten
Specifications
1.5 Lm
1.5 m
HBQ
1.5 Lm
∼230V Voltage
Very Flexible Heating Tape