Learn More
Masterflex™ HDPE Male Pipe Adapter Barbed Fitting
missing translation for 'orderingAttributeHoverText'
For Use With:
1/2 NPT(M) x 1/2 in. tubing I.D.
1/2 NPT(M) x 3/8 in. tubing I.D.
1/4 NPT(M) x 1/4 in. tubing I.D.
1/8 NPT(M) x 3/8 in. tubing I.D.
3/4 NPT(M) x 1/4 in. tubing I.D.
3/4 NPT(M) x 3/8 in. tubing I.D.
3/4 NPT(M) x 5/8 in. tubing I.D.
3/8 NPT(M) x 1/2 in. tubing I.D.
3/8 NPT(M) x 1/4 in. tubing I.D.
3/8 NPT(M) x 3/8 in. ID Tubing
Thread Size:
1/2 NPT(M)
1/4 NPT(M)
1/8 NPT(M)
3/4 NPT(M)
3/8 NPT(M)
missing translation for 'unitSize'
Pack of 10
Description
- Slip flexible tubing or hose onto the barb to create a seal that typically holds pressures up to 30 psi (2 bar)
- Available in a variety of configurations to meet your needs
- Adapts from a NPT male thread to a hose barb
- Sterilization through ethylyene oxide only
- Provided in packs of 10
Specifications
Specifications
| Material | HDPE |
| For Use With | 3/4 NPT(M) x 3/8 in. tubing I.D. |
| Packaging Quantity | 10 Pack |
| Sterilization Method | Ethylene Oxide |
| Thread Size | 3/4 NPT(M) |
| Shape | Straight |
| Product Type | Barbed Fitting |