Learn More
Buerkle™ Narrow-necked square bottle
Product Code. p-8002227 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
20 mL
50 mL
100 mL
Unit Size:
Each
Product Code. 10785164
Brand: Buerkle™ 03310020
Only null left
Add to basket
Add to basket
Cost-effective square bottle for packing and storing of liquids, powders and granules.
  • HDPE, transparent, shatterproof
  • Special labelling area for labels or manual lettering
  • Leakproof
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 20 mL
Color Transparent
Diameter (Metric) Inner Neck 13 mm
Material HDPE
Length (Metric) 27 mm
Quantity 1
Width (Metric) 27 mm
Thread Size (Metric) 18 mm
Closure Included Yes
Mouth Narrow
Height (Metric) 51 mm