Learn More
Huenersdorf™ HDPE Scoop
Product Code. 11712015
Click to view available options
Capacity (Metric):
250 mL
500 mL
725 mL
1500 mL
Unit Size:
Pack of 12
Pack of 3
Pack of 6
Product Code. 11712015
Brand: Huenersdorf™ 920600
Only null left
Add to basket
Add to basket
Use the Huenersdorf™ HDPE Scoop in the commercial sector, household and hobby.
  • Natural
  • Heavy quality
  • Sturdy HDPE material
  • Natural
  • Heavy quality
  • Sturdy HDPE material
Specifications
Capacity (Metric) 250 mL
Color Natural
Product Type Scoop
Includes 12/Pk.
Material HDPE
Length (Metric) 250 mm