Learn More
Huenersdorf™ HDPE Specimen Containers
Secure your items securely in Heunersdorf™ HDPE Specimen Containers, which are made of food-safe polypropylene (PP) for assorted uses.
Brand: Huenersdorf™ 910100
Additional Details : Weight : 1.19700kg
Description
- Resistant to acids and chemicals
- Tightly closing lid
- Deep freezable
Specifications
500 mL | |
LDPE | |
Natural | |
Square | |
Deep Freezer, Dishwasher, Microwave | |
Stackable | |
20 Pack | |
103 mm |
Natural | |
Hooded Lid | |
Polyethylene | |
Food Safe | |
Tightly Closing LDPE Lid | |
Specimen Container | |
103 mm | |
64 mm |