Learn More
Huenersdorf™ HDPE Specimen Containers
Secure your items securely in Heunersdorf™ HDPE Specimen Containers, which are made of food-safe polypropylene (PP) for assorted uses.
Brand: Huenersdorf™ 911600
Additional Details : Weight : 4.14000kg
Description
- Resistant to acids and chemicals
- Tightly closing lid
- Deep freezable
Specifications
2000 mL | |
LDPE | |
Natural | |
Round | |
Deep Freezer, Dishwasher, Microwave | |
Stackable | |
25 Pack | |
169 mm |
Natural | |
Hooded Lid | |
Polyethylene | |
Food Safe | |
Closure | |
Specimen Container | |
143 mm |