Learn More
BRAND™ HDPE Storage Bottle
Click to view available options
Capacity:
5 L
10 L
Description
- Constructed of high density polyethylene
- With screw cap and carrying handle
Specifications
Capacity | 5 L |
Includes | Carrying Handle |
Material | HDPE |
Type | Storage bottle |
Closure Included | Yes |
Closure Type | Screw Cap |
Mouth | Narrow |