Learn More
BRAND™ HDPE Storage Bottle
Shop All Brand Products
Click to view available options
Capacity (Metric):
5 L
10 L
Unit Size:
Each
Description
- Constructed of high density polyethylene
- With screw cap and carrying handle
Specifications
Specifications
| Capacity (Metric) | 10 L |
| Includes | Carrying Handle |
| Material | HDPE |
| Closure Included | Yes |
| Mouth | Narrow |
| Product Type | Storage Bottle |
| Closure Type | Screw Cap |