Learn More
Eppendorf™ Heat Sealer S200
Provides user-friendly and reliable hermetic sealing of a variety of multiwell plates.
4450.00€
Specifications
Hertz | 50/60 Hz |
---|---|
Voltage | 100/230 V |
Dimensions (L x W x H) | 220 × 321 × 425 mm |
Electrical Requirements | 100/230 V, 50/60 Hz |
For Use With (Equipment) | 96- and 384-well plates |
Description
- Eliminates evaporation in PCR
- Adjustable sealing temperatures for more flexibility
- Seals 96- and 384-well plates
- Sealing time selectable from 1 to 9 seconds in 0.5 second increments
- Integrated thermostat prevents overheating
- Powerful handle for ergonomic operation
- PCR plate adapter included
Specifications
50/60 Hz | |
220 × 321 × 425 mm | |
96- and 384-well plates |
100/230 V | |
100/230 V, 50/60 Hz | |
125°C to 200°C in 1°C increments |