Learn More
Heathrow Scientific™ Totemax™ Blood Collection Tray
Shop All Thermo Scientific Incubateurs à CO2 Products
Click to view available options
Color:
Blue
Green
Pink
Purple
Red
Unit Size:
Each
Description
- Made from food-grade HDPE for a durable non-breakable design
- Deep tray design helps reduce spills
- Hooks lock in place when not in use
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | Purple |
| Dimensions (L x W x H) | 43.2 x 27.9 x 27.9 mm (17 x 11 x 11 in.) |
| For Use With (Application) | Sample and Blood Collection |
| Height (English) | 11 in. |
| Height (Metric) | 27.9 mm |
| Length (English) | 17 in. |
| Length (Metric) | 43.2 mm |
| Size | 43.2 x 27.9 x 27.9 mm (17 x 11 x 11 in.) |
| Weight (Metric) | 27.9 mm |
| Show More |