Learn More
Fistreem International™ Heating Element, Calypso Ultrapure Water Still
Click to view available options
For Use With (Equipment):
2 Liter Model
4 Liter Model
Voltage:
220 V
240 V
Wattage:
0.9 kw
1.5 kw
Unit Size:
Each
Description
- A high temperature cut-off protects the heaters in the event of water failure while operator safety is assured through totally enclosed glassware.
- The Calypso Still is economical and available with 2L/hr or 4L/hr outputs.
Specifications
Specifications
| For Use With (Equipment) | 2 Liter Model |
| Type | Heating Element |
| For Use With (Application) | Calypso Ultrapure Water Still |
| Voltage | 240 V |
| Wattage | 0.9 kw |