Learn More
Rogo Sampaic™ Heavy Crystallizing Dishes
Product Code. p-8008546
Click to view available options
Capacity (Metric):
1 L
2 L
9 L
Diameter (Metric):
164 mm
205 mm
335 mm
Height (Metric):
78 mm
96 mm
145 mm
Unit Size:
Each
Pack of 3
Pack of 5
Product Code. 11714156
Brand: Rogo Sampaic™ CNN004
Only null left
Add to basket
Add to basket
Ideal for crystallization needs in the laboratory. Rogo Sampaic™ Heavy Crystallizing Dishes are manufactured from clear soda lime glass.
  • Also excellent for storage
  • Available in a range of sizes
Specifications
Capacity (Metric) 1 L
Type Heavy Crystallizing Dish
Height (Metric) 78 mm
Diameter (Metric) 164 mm