Learn More
Hecht Karl™ Heavy-wall Test Tubes
Manufactured with AR-Glas™ (soda glass) to minimize breakage. Karl Hecht™ Heavy-Wall Test Tubes are suitable for general laboratory use.
Brand: Hecht Karl™ 42775051
Additional Details : Weight : 1.54000kg
Description
- Soda lime glass
- Round bottom
- Rimless
- Sized to support all laboratory needs
Specifications
Round Bottom | |
160 mm | |
100 Pack | |
15.25 mm |
Test tube | |
Soda Glass | |
No |