Learn More
Ansell HyFlex™ 70-225 Medium Duty String Knit Gloves with Aramid Fiber
Product Code. 12666921 Shop All Ansell Products
Click to view available options
:
9
Product Code. 12666921
Brand: Ansell 70225090
Only null left
Add to basket
Add to basket
Aramid fiber medium duty industrial gloves offers high cut-resistance to weight ratio. Ideal for automotive or machinery and equipment industries.
  • Five times the cut resistance of leather
  • Heat protection
  • Hand-to-hand reversible
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Neptune
Color Yellow
Cut Resistance Rating ANSI A3
Hand Ambidextrous
Latex Free Yes
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size 9
Glove Cuff Style Knit