Learn More
Heidolph™ Hei-VAP Core ML G6, EU-plug
Product Code. 17323300 Shop All Heidolph Instruments Products
Click to view available options
Capacity (Metric):
5 L
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 17323300
Brand: Heidolph™ 5720160000
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
The Hei-VAP Core ML G6 is focused on the essentials: a clear digital display with access to the basic parameters at any time.
An operating concept that is focused on the essentials: clear and with access to the basic parameters rotation speed and heating bath temperature at any time.
  • Clear digital display showing actual and set values
  • Two separate knobs with dynamic control for setting the rotation speed and heating bath temperature: fast turning for large jumps, slow turning for precise setting of the set values
  • Accidental changing of the values can be prevented by the lock function
  • Intuitive LED ring light system makes activated functions clearly visible even from a distance
  • Easy adjustment of the immersion depth up to 155 mm and the inclination angle from 20° to 80°
  • Standby function and residual heat warning at heating bath temperatures above 50°C
  • Motor lift model with G6 vertical glassware for reflux distillation
  • The standby button stops all functions and, when using a motor lift model, lifts the evaporating flask from the heating bath
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 5 L
Type Motor Lift Rotary Evaporator
Plug Type EU-plug