Learn More
Heidolph™ Hei-VAP Powder Drying Package, EU-plug
Product Code. 17393330 Shop All Heidolph Instruments Products
Click to view available options
Plug Type:
EU-plug
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 17393330
Brand: Heidolph™ 5743230000
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
The Hei-VAP Powder Drying Package features a motor lift, powder flask, and tubing set.
  • Hei-VAP Expert Control with motor lift and vertical glassware set G3
  • Vacuum pump Rotavac Valve Control
  • Vacuum valve
  • Tubing set
  • Powder flask 2 L
  • Vapor tube with powder filter
TRUSTED_SUSTAINABILITY
Specifications
Type Motor Lift Rotary Evaporator
Plug Type EU-plug