Learn More
Horst cartridge heater, DxL = 8 x 50mm, 125W
Brand:  Horst 512050
cartridge heater, DxL = 8 x 50mm, 125W
Product Code. 10007021
  • ONLINE EXCLUSIVE PRICE
    61.53€
    54.02€ / Each
    Save 7.51€
    12% Off
Only null left
Estimated Shipment: 03-02-2026