Learn More
Helios™ Helios Italquartz™ Evaporating Dishes
Click to view available options
Capacity (Metric):
20 mL
55 mL
Diameter (Metric):
47 mm
61 mm
77 mm
Height (Metric):
15 mm
18 mm
27 mm
Unit Size:
Each
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 20 mL |
| Material | Quartz |
| Shape | Round |
| Type | Evaporating Dish |
| Height (Metric) | 15 mm |
| Diameter (Metric) | 47 mm |