missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES, 0.5M buffer soln., pH 8.0

Catalog Number p-7042572
Click to view available options
Quantity:
100 mL
250 mL
7365-45-9
C8H18N2O4S
238.30
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
CHEBI:42334
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
OCCN1CCN(CCS(O)(=O)=O)CC1
7365-45-9
C8H18N2O4S
238.30
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
CHEBI:42334
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
OCCN1CCN(CCS(O)(=O)=O)CC1

HEPES has been utilized extensively for research and commercial applications in the cell culture and tissue culture industries. It is useful in the pH range 6.8-8.2. HEPES is used as a zwitterionic, all-purpose “Good’s” buffer for use in biological research. HEPES is widely used in cell culture, largely because it is better at maintaining physiological pH despite changes in carbon dioxide concentration when compared to bicarbonate buffers, which are also commonly used in cell culture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES has been utilized extensively for research and commercial applications in the cell culture and tissue culture industries. It is useful in the pH range 6.8-8.2. HEPES is used as a zwitterionic, all-purpose “Good’s” buffer for use in biological research. HEPES is widely used in cell culture, largely because it is better at maintaining physiological pH despite changes in carbon dioxide concentration when compared to bicarbonate buffers, which are also commonly used in cell culture.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Concentration 0.5 M
pH 8.0
Physical Form Liquid
Quantity 100 mL
Solubility Information Fully miscible.
Chemical Name or Material HEPES buffer soln.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'