Learn More
Thermo Scientific™ HEPES Lysis Buffer with Triton™ X-100, 2X
Product Code. 15485109
Click to view available options
Quantity:
125mL
250mL
Unit Size:
125mL
250mL
Product Code. 15485109
Brand: Thermo Scientific™ J62809.AK
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 250mL
Format Liquid
Product Type Cell Lysis Reagent
Concentration 2X
Content And Storage Ambient temperatures