missing translation for 'onlineSavingsMsg'
Learn More

Heptane, Puriss. p.a., Reag. Ph. Eur., ≥99% n-heptane basis (GC), Honeywell Riedel-de Haën™

Brand:  Honeywell Riedel-de Haën 32287-1L

Additional Details : CAS : 142-82-5 Poids : 0.68400kg Transport : UN number : 1206 Chem class : 3 Pack group : II


Quantity 1L

 View more versions of this product

Product Code. 15653750

  • 75.40€ / 1 L


En stock, prêt à être expédié : 51
Add to basket

Description

Description

Specifications

Chemical Identifiers

142-82-5
100.205
IMNFDUFMRHMDMM-UHFFFAOYSA-N
8900
heptane
C7H16
MFCD00009544
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CHEBI:43098
CCCCCCC

Specifications

Heptane
142-82-5
CH3(CH2)5CH3
1206
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CCCCCCC
100.205
CHEBI:43098
≥99% n to heptane basis (GC)
0.684g/mL at 25°C
433°F
3.5 (vs air)
1L
Reag. Ph. Eur.
C7H16
MFCD00009544
1730763
IMNFDUFMRHMDMM-UHFFFAOYSA-N
heptane
8900
100.2g/mol
puriss. p.a.
0.07
40mmHg (20°C), 83mmHg (37.7°C)
−4°C (24.8°F)
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • n-Heptane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS Documents