Learn More
Notrax Heritage Rib™ Economical Ribbed Needle-Punched Entrance Mat
Economical ribbed needle-punched carpet mats making them highly effective entrance mat products. Durable fibers offer good scraping action, preventing moisture and grime from being walked into facilities.
Brand:  Notrax 117S0023CH
Additional Details : Weight : 1.70000kg
 View more versions of this product
Product Code. 17711928
  • 26.10€ / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket
Description
Description
  • This is an economical yet highly effective entrance mat product
  • Durable fibers offer good scraping action, preventing moisture and grime from being walked into facilities
  • Low profile allows foot, cart and wheelchair traffic to move easily over the mat
  • Vinyl backing is moisture resistant and slip resistant
Specifications
Specifications
35 in.
Industrial and Commercial Matting
60 cm
Charcoal
Polypropylene Fibers
90 cm