Learn More
Notrax Heritage Rib™ Economical Ribbed Needle-Punched Entrance Mat
Economical ribbed needle-punched carpet mats making them highly effective entrance mat products. Durable fibers offer good scraping action, preventing moisture and grime from being walked into facilities.
Brand: Notrax 117S0035CH
Additional Details : Weight : 4.20000kg
Description
- This is an economical yet highly effective entrance mat product
- Durable fibers offer good scraping action, preventing moisture and grime from being walked into facilities
- Low profile allows foot, cart and wheelchair traffic to move easily over the mat
- Vinyl backing is moisture resistant and slip resistant
Specifications
59 in. | |
Industrial and Commercial Matting | |
90 cm |
Charcoal | |
Polypropylene Fibers | |
150 cm |