missing translation for 'onlineSavingsMsg'
Learn More

Hexanoic acid-ethyl ester 10 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30313968
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30313968 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 30313968

Brand: Dr. Ehrenstorfer DREA14196200AL10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Caproic acid ethyl ester
CAS 123-66-0
Analyte or Component Names Caproic acid ethyl ester
Shipping Condition Room Temperature
Molecular Formula C8 H16 O2
Concentration 10 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C8H16O2/c1-3-5-6-7-8(9)10-4-2/h3-7H2,1-2H3
SMILES CCCCCC(=O)OCC
IUPAC Name ethyl hexanoate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 144.21
Quantity 1 mL
Formula Weight 144.115 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.