Learn More
Hexanophenone, 98%, ACROS Organics™
Brand: Acros Organics 120721000
Additional Details : CAS : 942-92-7 Poids : 0.10000kg
Packaging | Glass bottle |
---|---|
Quantity | 100g |
Chemical Identifiers
942-92-7 | |
176.26 | |
MAHPVQDVMLWUAG-UHFFFAOYSA-N | |
70337 | |
CCCCCC(=O)C1=CC=CC=C1 |
C12H16O | |
MFCD00009512 | |
hexanophenone, caprophenone, 1-hexanone, 1-phenyl, 1-phenyl-1-hexanone, n-hexanophenone, amyl phenyl ketone, hexaphenone, pentyl phenyl ketone, 4-ethylbutyrophenone, n-amylphenyl ketone | |
1-phenylhexan-1-one |
Specifications
Hexanophenone | |
>112°C | |
Glass bottle | |
0.95 | |
Yellow | |
100g | |
942-92-7 | |
C12H16O | |
MFCD00009512 | |
hexanophenone, caprophenone, 1-hexanone, 1-phenyl, 1-phenyl-1-hexanone, n-hexanophenone, amyl phenyl ketone, hexaphenone, pentyl phenyl ketone, 4-ethylbutyrophenone, n-amylphenyl ketone | |
CCCCCC(=O)C1=CC=CC=C1 | |
176.26 | |
176.26 | |
98% |
0.9500g/mL | |
Authentic | |
1.5105 | |
265.1°C | |
25.0°C to 26.0°C | |
98% | |
98% | |
C6H5CO(CH2)4CH3 | |
07,II,257 | |
MAHPVQDVMLWUAG-UHFFFAOYSA-N | |
1-phenylhexan-1-one | |
70337 | |
Liquid After Melting |