missing translation for 'onlineSavingsMsg'
Learn More

Hexanophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-142569
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
942-92-7
C12H16O
176.26
MFCD00009512
MAHPVQDVMLWUAG-UHFFFAOYSA-N
hexanophenone, caprophenone, 1-hexanone, 1-phenyl, 1-phenyl-1-hexanone, n-hexanophenone, amyl phenyl ketone, hexaphenone, pentyl phenyl ketone, 4-ethylbutyrophenone, n-amylphenyl ketone
70337
1-phenylhexan-1-one
CCCCCC(=O)C1=CC=CC=C1
This item is not returnable. View return policy
942-92-7
C12H16O
176.26
MFCD00009512
MAHPVQDVMLWUAG-UHFFFAOYSA-N
hexanophenone, caprophenone, 1-hexanone, 1-phenyl, 1-phenyl-1-hexanone, n-hexanophenone, amyl phenyl ketone, hexaphenone, pentyl phenyl ketone, 4-ethylbutyrophenone, n-amylphenyl ketone
70337
1-phenylhexan-1-one
CCCCCC(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 25.0°C to 26.0°C
Color Yellow
Density 0.9500g/mL
Boiling Point 265.1°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5CO(CH2)4CH3
Refractive Index 1.5105
Quantity 5 g
Beilstein 07,II,257
Specific Gravity 0.95
Formula Weight 176.26
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Hexanophenone, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'