missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexyl Chloroformate, TRC
Specifications
Specifications
| Chemical Name or Material | Hexyl Chloroformate |
| CAS | 6092-54-2 |
| Molecular Formula | C7H13ClO2 |
| Quantity | 1 g |
| Synonym | Carbonochloridic Acid Hexyl Ester, Chloroformic Acid Hexyl Ester, Hexyl Alcohol Chloroformate, Chloroformic Acid n-Hexyl Ester, Hexyl Chlorocarbonate, Hexyloxycarbonyl Chloride, n-Hexyl Chloroformate |
| InChI Formula | InChI=1S/C7H13ClO2/c1-2-3-4-5-6-10-7(8)9/h2-6H2,1H3 |
| SMILES | CCCCCCOC(=O)Cl |
| IUPAC Name | hexyl carbonochloridate |
| Molecular Weight (g/mol) | 164.63 |
| Formula Weight | 164.0604 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?