Learn More
Novus Biologicals™ HIF-1 alpha Knockout Hypoxic-treated/untreated HeLa Cell Lysate
Product Code. 18649266 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18649266 2 Vials Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18649266 Supplier Novus Biologicals™ Supplier No. NBP311827
Cell Lysate
TRUSTED_SUSTAINABILITY
Specifications
Preparation Method HIF-1 alpha KO HeLa Whole cell lysate is provided as a Western blot positive control for Hif-1 research. The lysates are provided as a set of hypoxic and normoxic samples. Use 10 μL / lane for a standard mini-gel blot (approx. 1 mg/mL).
Quantity 2 Vials
Host Species Human
Content And Storage Aliquot and store at -20°C to +80°C. Avoid freeze/thaw cycles.
Research Category Adaptive Immunity, Angiogenesis, Apoptosis, Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Cytokine Research, HIF Target Genes, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators
Target HIF-1 alpha
Synonym AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1 A, HIF-1 alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8
Gene Symbol HIF1A