missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals HIF2A / HIF2 alpha antibody 0,1ml

Brand:  Novus Biologicals NB100-122

Encompass

HIF2A / HIF2 alpha antibody 0,1ml

Product Code. 11395362

  • 569.10€ / 0.1mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.