Learn More
Heraeus™ High Form Platinum Crucible
Product Code. 11950008
Click to view available options
Capacity (Metric):
12 mL
15 mL
20 mL
25 mL
30 mL
40 mL
50 mL
Unit Size:
Each
Product Code. 11950008
Brand: Heraeus™ TI19PT11
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Use for various fusion or melting processes in the analytical lab. Heraeus™ High Form Platinum Crucible is a range of containers with good resistance to aggressive media and stability even at high temperatures. Crucibles use a dispersion hardened allot for increased strength and service times.
The use of precious metals is still absolutely essential because of their good chemical resistance to aggressive media. Precious metals remain stable even at high temperatures. The inclusion of finely dispersed zirconia, creating a Dispersion Hardened (DPH) allow, offers an economical way to increase the mechanical strength of the product resulting in longer service times.
  • High quality, according to established standards/norms
  • Competitive pricing
  • Crucible lids sold separately
  • Available in a range of sizes and working capacities
Specifications
Description Platinum, with dispersed zirconia; Smart Standard: Ti 1/9
Diameter (Metric) Inner Top: 40mm, Bottom: 25mm
Material Platinum
For Use With (Application) Fusion or melting processes
Quantity 1 Each
Capacity (Metric) 40 mL
Height (Metric) 45mm
Shape Tall Form