Learn More
Heraeus™ High Form Platinum Crucible
Click to view available options
Capacity (Metric):
12 mL
15 mL
20 mL
25 mL
30 mL
40 mL
50 mL
Unit Size:
Each
Description
The use of precious metals is still absolutely essential because of their good chemical resistance to aggressive media. Precious metals remain stable even at high temperatures. The inclusion of finely dispersed zirconia, creating a Dispersion Hardened (DPH) allow, offers an economical way to increase the mechanical strength of the product resulting in longer service times.
- High quality, according to established standards/norms
- Competitive pricing
- Crucible lids sold separately
- Available in a range of sizes and working capacities
Specifications
Specifications
| Description | Platinum, with dispersed zirconia; Smart Standard: Ti 1/8 |
| Diameter (Metric) Inner | Top: 36mm, Bottom: 22mm |
| Material | Platinum |
| For Use With (Application) | Fusion or melting processes |
| Quantity | 1 Each |
| Capacity (Metric) | 30 mL |
| Height (Metric) | 40mm |
| Shape | Tall Form |