Learn More
Abeba™ High Original 5009 Shoes
ESD dissipative shoe includes PU sole with high slip resistance.
Brand:  Abeba™ 5009/39
Additional Details : Poids : 0.55000kg
 View more versions of this product
Product Code. 15651778
  • 37.30€ / 1 pair
Expédition estimée
01-10-2020
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather, perforated
  • Black
  • Slip resistance PU outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Leather top layer
  • Narrow fit
  • Wedge heel 45mm
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Leather
Slip-resistant
5009
Black
39
Polyurethane