Learn More
Techne™ Temperature Bath Fluid
Enhance the performance of your Techne baths and circulators
Brand:  Techne™ 12122-99
Additional Details : Weight : 0.10000kg
Product Code. 15197036
  • 1070.00€ / Each
Estimated Shipment: 01-07-2024
to see stock.
Add to basket
Description
Description
  • Enhances bath temperature uniformity and stability
  • Safe and non-toxic high temperature bath fluid that is versatile and cost effective
Specifications
Specifications
18 L
High Temperature Heat Transfer Fluid
Synthetic Alkylated Aromatics
315°C
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Dynalene SF (hydrocarbon mixture)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P264-Wash skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362-Take off contaminated clothing.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : benzene, mono-C12-14-alkyl derivs.