Learn More
Nabertherm™ High-Temperature Ovens, Forced Convection Chamber Furnaces
Product Code. 17308059 Shop All Nabertherm Products
Click to view available options
Capacity (Metric):
30 L
60 L
120 L
250 L
500 L
Controller Type:
B400
B500
B510
C540
C550
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 17308059
Brand: Nabertherm™ NA1283M2
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
Chamber furnaces with air circulation are characterized by their extremely high temperature uniformity. Suitable for processes such as annealing, crystalizing, preheating, curing, and tool making.
  • Horizontal air circulation with optimum distribution through stainless steel baffles
  • Swing door hinged on the right
  • Temperature uniformity up to +/- 4 C according to DIN 17052-1 (NAT 15/65 and NAT 30/85 up to +/- 5°C) in the empty work space
  • One frame sheet and rails for two additional trays included in the scope of delivery (NAT 15/65 and NAT 30/85 without frame sheet)
  • Base frame included in the delivery, NAT 15/65 and NAT 30/85 designed as table-top model
Specifications
Controller Type C540
Capacity (Metric) 120 L
Temperature (Metric) 850°C
Dimensions (D x W x H) Interior 600 x 450 x 450 mm
Dimensions (L x W x H) Exterior 1420 x 890 x 1540 mm
Voltage 400 V
Wattage 14000 W