Learn More
Nabertherm™ High-Temperature Ovens, Forced Convection Chamber Furnaces
Product Code. 17348049 Shop All Nabertherm Products
Click to view available options
Capacity (Metric):
30 L
60 L
120 L
250 L
500 L
Controller Type:
B400
B500
B510
C540
C550
Unit Size:
Each
Product Code. 17348049
Brand: Nabertherm™ NA0683M2
Only null left
Add to basket
Add to basket
Chamber furnaces with air circulation are characterized by their extremely high temperature uniformity. Suitable for processes such as annealing, crystalizing, preheating, curing, and tool making.
  • Horizontal air circulation with optimum distribution through stainless steel baffles
  • Swing door hinged on the right
  • Temperature uniformity up to +/- 4 C according to DIN 17052-1 (NAT 15/65 and NAT 30/85 up to +/- 5°C) in the empty work space
  • One frame sheet and rails for two additional trays included in the scope of delivery (NAT 15/65 and NAT 30/85 without frame sheet)
  • Base frame included in the delivery, NAT 15/65 and NAT 30/85 designed as table-top model
Specifications
Controller Type C540
Capacity (Metric) 60 L
Temperature (Metric) 850°C
Dimensions (D x W x H) Interior 500 x 350 x 350 mm
Dimensions (L x W x H) Exterior 1330 x 790 x 1440 mm
Voltage 400 V
Wattage 11000 W