Learn More
Cytiva His MultiTrap™ TALON™
Shop All Cytiva Products
missing translation for 'orderingAttributeHoverText'
Quantity:
4/Pk.
missing translation for 'unitSize'
Each
Beschreibung
Prepacked 96-well plate for expression screening and small-scale parallel purification of histidine-tagged proteins
Specifications:
- Ligand - TALON Superflow Cobalt-IMAC
- Matrix - Cross-linked agarose, 6%
- Particle size - 60 to 160μm
- Binding Capacity/mL - Up to 20 mg histidine-tagged protein
- pH stability working range - 3 to 12
- No of wells: 96
- Material - Polypropylene (PP) and polyethylene (PE)
- Dimensions - 127.8 x 85.5 x 30.6 mm
- Complete packsize - 4 x 96-well filter plates
- Volume - 50μL
Spezifikation
Spezifikation
| Quantity | 4/Pk. |
| For Use With (Application) | For High Throughput Screening of Histidine-Tagged Proteins From Clarified Cell Lysates |
| Storage Requirements | 4°C to 8°C |
| Binding Capacity | Up to 20 mg Histidine-tagged Protein |
| Product Type | His MultiTrap Talon prepacked Column |
| Particle Size | 60 to 160 μm |
| Material | Polypropylene, Polyethylene |
| Matrix | 6% Cross-Linked Agarose |
| pH Range | 3 to 12 |
| Ligand Type | Talon Superflow Cobalt-IMAC |
Sicherheit und Handhabung
Signalwort
- Warning
- Flammable liquid Category 3
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur