Learn More
Thermo Scientific™ HisPur™ Cobalt Chromatography Cartridges, 1 mL
Gently purify His-tagged recombinant fusion proteins using cobalt (Co2+) immobilized metal chelate affinity (IMAC) beads, spin columns and plates.
234.00€ - 413.00€
Specifications
Content And Storage | Store between 4°C and 8°C. |
---|---|
Form | Liquid Suspension |
Product Line | HisPur™ |
Target | His-Tagged Protein |
Type | Chromatography Cartridge |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
11804211
|
Thermo Scientific™
90093 |
HisPur Cobalt Chromatography Cartridges, 1mL | 5 Cartridges |
234.00€
Pack of 5 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
11814211
|
Thermo Scientific™
90094 |
HisPur Cobalt Chromatography Cartridges, 5mL | 2 Cartridges |
413.00€
Pack of 2 |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Thermo Scientific™ HisPur Cobalt Resin is an immobilized metal chelate affinity (IMAC) support for specific, fast and gentle His-tagged purification.
Additional Information and Online Ordering »
Highlights:
High purity – obtain more than 10 milligrams of pure His-tagged protein per milliliter of resin without optimizing imidazole washing conditions
Low metal leaching – no metal contamination in eluted histidine-tagged protein sample
Versatility – purify proteins under native or denaturing conditions; compatible with cell lysis reagents and a variety of buffer additives
Flexibility – available as bulk resin or predispensed columns, chromatography columns and 96-well filter plates
Cost effective – resin can be reused several times or discarded after single use
Superior – performs better than other commercially available IMAC Resins
Available Formats:
- Resin Slurries – crosslinked 6% beaded agarose; 10mL, 100mL, 500mL bottles
- Spin Columns – 0.2mL (microcentrifuge), 1mL, 3mL columns
- Purification Kits – complete kits in all three column sizes
- Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
- 96-well Spin Plates – pre-dispensed 96-well filter plates, compatible with centrifugation and vacuum manifold systems for manual or automated purification. The plates enable fast, consistent well-to-well and plate-to-plate reproducibility for small-scale, high-throughput separations
- Bulk and Custom – let us know your needs for bulk HisPur Cobalt Resin
Includes:
Kits contain HisPur Cobalt Columns, equilibration/wash buffer and elution buffer
Specifications
Store between 4°C and 8°C. | |
HisPur™ | |
Chromatography Cartridge | |
Affinity, Agarose Resin |
Liquid Suspension | |
His-Tagged Protein | |
Metal Chelate, Divalent Cobalt |
For Research Use Only. Not for use in diagnostic procedures.