missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HJ 827-2017 Carbamate Pesticides Mixture 154 50-1000 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2,3,5-Trimethacarb∣Bendiocarb∣Carbaryl∣Carbofuran∣Carbofuran-3-hydroxy∣Fenobucarb∣Isoprocarb∣Methiocarb∣Methomyl∣Methomyl-oxime∣Metolcarb∣Pirimicarb∣Promecarb∣Propoxur |
| CAS | 2655-15-4, 22781-23-3, 63-25-2, 1563-66-2, 16655-82-6, 3766-81-2 |
| Analyte or Component Names | 2,3,5-Trimethacarb∣Bendiocarb∣Carbaryl∣Carbofuran∣Carbofuran-3-hydroxy∣Fenobucarb∣Isoprocarb∣Methiocarb∣Methomyl∣Methomyl-oxime∣Metolcarb∣Pirimicarb∣Promecarb∣Propoxur |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Carbofuran 50 μg/mL∣Isoprocarb 50 μg/mL∣Methiocarb 50 μg/mL∣Pirimicarb 50 μg/mL∣Fenobucarb 100 μg/mL∣Metolcarb 100 μg/mL∣Promecarb 100 μg/mL∣2,3,5-Trimethacarb 200 μg/mL∣Bendiocarb 200 μg/mL∣Carbaryl 200 μg/mL∣Carbofuran-3-hydroxy 200 μg/mL∣Methomyl 200 μg/mL∣Propoxur 200 μg/mL∣Methomyl-oxime 1000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?