missing translation for 'onlineSavingsMsg'
Learn More

HLA A Antibody Pair, Novus Biologicals™

Product Code. 18362518 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362518 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18362518 Brand Novus Biologicals Brand No. H00003105AP21

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody Pair

HLA A Polyclonal antibody specifically detects HLA A in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA A
Applications Sandwich ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation 1x PBS, pH 7.4
Gene Alias A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80
Quantity 1 Set
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3105
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.