Learn More
Hoefer™ Blotting Paper
Shop All Cytiva Products
Click to view available options
Length (Metric):
9 cm
14 cm
14.5 cm
21 cm
Width (Metric):
10.5 cm
16 cm
21.5 cm
25 cm
For Use With (Equipment):
TE22 Mini Tank or MiniVE Module
TE42 or TE62 Tank Blotting Units
TE70 or TE77 Semi-Dry Blotting Unit
TE77 Semi-Dry Blotting Unit
Unit Size:
Pack of 25
Pack of 50
Description
- For Western Blotting
Specifications
Specifications
| Length (Metric) | 14 cm |
| Width (Metric) | 16 cm |
| For Use With (Equipment) | TE70 or TE77 Semi-Dry Blotting Unit |
| Format | Sheet |
| Material | Cellulose |