Learn More
Hoefer™ Combs for SUB25 Maxi-Plus Submarine Gel Electrophoresis Unit
For use with SUB25 and SUB25C Units
87.35€
Specifications
For Use With (Application) 52 sample throughput (multi-channel pipette compatible)
For Use With (Equipment) SUB25 Maxi-Plus Unit
Width (English) Gel 9.84 in.
View More Specs
Products 4
Product Code Brand Thickness (English) Thickness (Metric) Volume (English) Volume (Metric) Price Quantity & Availability  
Product Code Brand Thickness (English) Thickness (Metric) Volume (English) Volume (Metric) Price Quantity & Availability  
12685786
View Documents Promotion Details
Hoefer™
SUB25-C1.5-52MC
0.05 in. 1.5 mm 0.0006 oz. 20 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12605796
View Documents Promotion Details
Hoefer™
SUB25-C1-52MC
0.03 in. 1 mm 0.0004 oz. 13 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12625796
View Documents Promotion Details
Hoefer™
SUB25-C2-52MC
0.07 in. 2 mm 0.0008 oz. 25 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12675786
View Documents Promotion Details
Hoefer™
SUB25-C1.5-26MC
0.05 in. 1.5 mm 0.001 oz. 45 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description
Description
  • Either 26 or 52 Wells
  • 1, 1.5 and 2 mm Thickness
  • 3 or 7 mm Well Width
  • Min. 13, max. 60 μL Sample Volume
Specifications
Specifications
52 sample throughput (multi-channel pipette compatible)
SUB25 Maxi-Plus Unit
9.84 in.