Learn More
Hoefer™ Combs for SUB25 Maxi-Plus Submarine Gel Electrophoresis Unit
For use with SUB25 and SUB25C Units
Brand:  Hoefer™ SUB25-C2-52MC
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12625796
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Either 26 or 52 Wells
  • 1, 1.5 and 2 mm Thickness
  • 3 or 7 mm Well Width
  • Min. 13, max. 60 μL Sample Volume
Specifications
Specifications
52 sample throughput (multi-channel pipette compatible)
0.07 in.
0.0008 oz.
9.84 in.
SUB25 Maxi-Plus Unit
2 mm
25 μL