missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ HE-PLUS Casting Kits
For use with HE-PLUS Horizontal Electrophoresis System
390.00€
Specifications
For Use With (Equipment) | HE-plus Horizontal Electrophoresis System |
---|
Includes: Mini Casting Kit includes four gel trays (6 × 6cm), two combs (5/8 wells) and casting stand. Standard Casting Kit includes one large gel tray (12.5 × 13cm), two medium gel trays (12.5 × 6cm), four combs (14/28 wells) and casting stand.
Product Code | Brand | Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Includes | Price | Quantity & Availability | ||||
15348693
|
Hoefer™
HE-PLUS16 |
Mini Casting Kit | Four gel trays (6 x 6cm), two combs (5/8-well) and casting stand |
390.00€
1 set |
In Stock
Log in to see stock. |
||||
15378693
|
Hoefer™
HE-PLUS18 |
Standard Casting Kit | One large gel tray (12.5 x 13cm), two medium gel trays (12.5 x 6cm), four combs (14/28-well) and casting stand |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
Description
- Accessories and Replacement Parts for HE Systems
Specifications
HE-plus Horizontal Electrophoresis System |