Learn More
Hoefer™ Mini and Standard Electrophoresis Blotting Systems
Product Code. 11945388
Click to view available options
Includes:
Mini Blot Unit with Semidry Unit
Mini Blot Unit with Transfer Tank
Std Blot Unit with Semidry Unit
Std Blot Unit with Transfer Tank
Unit Size:
Each
Product Code. 11945388
Brand: Hoefer™ 326703
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Includes
The Mini Electrophoresis Blotting System with Transfer Tank includes the Mighty Small II Mini Vertical Electrophoresis System, Model SE260 Complete; TE22 Mini Tank Blotting Unit; and PS300B Power Supply (300V, 500mA, 90w). The Mini Electrophoresis Blotting System with Semidry Unit includes the Mighty Small II Mini Vertical Electrophoresis System, Model SE260 Complete; Semidry Transfer Unit with Built-In Power Supply, Model TE70XP; and PS300B Power Supply (300V, 500mA, 90w). The Standard Electrophoresis Blotting System with Transfer Tank includes the SE600 Chroma Vertical Electrophoresis System; TE62 Tank Blotting Unit; and PS300B Power Supply (300V, 500mA, 90w). The Standard Electrophoresis Blotting System with Semidry Unit includes the SE600 Chroma Vertical Electrophoresis System; Large Semidry Transfer Unit with Built-In Power Supply, Model TE77XP; and PS300B Power Supply (300V, 500mA, 90w).
Economical equipment packages provide proven electrophoresis solutions for Western blotting
  • Comes with PS300B 300 V, 500 mA, 90 W Power Supply for all units
  • Save time and money with a combination package
Specifications
Content And Storage Mighty Small™ II mini deluxe vertical unit, semi dry transfer unit with built-in power supply
Type Mini Electrophoresis Blotting System
Includes Mini Blot Unit with Semidry Unit
Voltage 300 V
Wattage 90 w
Amperage 500 mA