Learn More
Hoefer™ Mini and Standard Electrophoresis Blotting Systems
Description
Includes
The Mini Electrophoresis Blotting System with Transfer Tank includes the Mighty Small II Mini Vertical Electrophoresis System, Model SE260 Complete; TE22 Mini Tank Blotting Unit; and PS300B Power Supply (300V, 500mA, 90w). The Mini Electrophoresis Blotting System with Semidry Unit includes the Mighty Small II Mini Vertical Electrophoresis System, Model SE260 Complete; Semidry Transfer Unit with Built-In Power Supply, Model TE70XP; and PS300B Power Supply (300V, 500mA, 90w). The Standard Electrophoresis Blotting System with Transfer Tank includes the SE600 Chroma Vertical Electrophoresis System; TE62 Tank Blotting Unit; and PS300B Power Supply (300V, 500mA, 90w). The Standard Electrophoresis Blotting System with Semidry Unit includes the SE600 Chroma Vertical Electrophoresis System; Large Semidry Transfer Unit with Built-In Power Supply, Model TE77XP; and PS300B Power Supply (300V, 500mA, 90w).
- Comes with PS300B 300 V, 500 mA, 90 W Power Supply for all units
- Save time and money with a combination package
Specifications
Specifications
| Content And Storage | Deluxe dual cooled vertical unit, large semi dry transfer unit with built-in power supply |
| Type | Standard Electrophoresis Blotting System |
| Includes | Std Blot Unit with Semidry Unit |
| Voltage | 300 V |
| Wattage | 90 w |
| Amperage | 500 mA |