Learn More
Hoefer™ miniVE Integrated Vertical Electrophoresis
Product Code. 15588971
Change view
Click to view available options
Dimensions (L x W x H):
32 x 37 x 14 cm
Unit Size:
1 set
Product Code. Dimensions (L x W x H) unitSize
15588971 32 x 37 x 14 cm 1 set
1 options
Product Code. 15588971
Brand: Hoefer™ SE300
Only null left
Add to basket
Add to basket
Perform both electrophoresis and electro transfer in one sturdy, compact unit.
  • Basic
Specifications
For Use With (Application) Protein electrophoresis, Electro transfer (blotting)
Humidity Range Up to 80%
Includes Lower Buffer Chamber (tank); Gel Modules with Gaskets, 2 assembliesl Safety Lid with High Voltage Leads; Well-Locating Decals, 2 pcs; Wonder Wedge Plate Separation Tool Gel Seal
Model Basic
Dimensions (L x W x H) 32 x 37 x 14 cm