Learn More
Hoefer™ SE 260 or miniVE Vertical Electrophoresis Unit Accessory, Plates
Use with SE 260 or miniVE vertical electrophoresis units. Hoefer™ SE 260 or miniVE Vertical Electrophoresis Unit Accessory, Plates come in a variety of styles for particular needs.
77.55€ - 201.00€
Specifications
For Use With (Equipment) | SE 260, MiniVE™ Vertical Electrophoresis Unit |
---|
Product Code | Brand | Type | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Material | Price | Quantity & Availability | ||||
11549693
|
Hoefer™
SE262P-5 |
Low Fluorescence Plate | Glass |
77.55€
Pack of 5 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
12094707
|
Hoefer™
SE262N-5 |
Notched Plate | Alumina |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12054657
|
Hoefer™
SE262GN-5 |
Low Fluorescence Notched Plate | Glass |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
Description
- Size: 10 x 10.5 cm
Specifications
SE 260, MiniVE™ Vertical Electrophoresis Unit |