missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ SUB10 Mini-Plus Submarine Combs

Accessories for SUB10 Mini-Plus Submarine

Brand:  Hoefer™ SUB10-C1-16

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12695716

  • 54.53€ / Each


Estimated Shipment
13-07-2021
Add to basket

Description

Description

Specifications

Specifications

0.19 in.
SUB10 Mini plus Submarine Unit
16
0.1 cm
0.14 in.
0.5 cm
Comb
0.04 in.
15 μL
0.36 cm
Videos
SDS
Documents