Learn More
Hoefer™ SUB13 Midi Submarine Combs
Click to view available options
No. of Wells:
10
12
16
20
24
28
Thickness (Metric):
0.1 cm
0.15 cm
0.2 cm
Width (Metric) Well:
0.3 cm
0.4 cm
0.55 cm
0.8 cm
0.95 cm
Unit Size:
Each
Description
- Ranges from 10, 12, 16, 20, 24 and 28 Wells
- 1, 1.5 and 2 mm Thickness
- 3 to 9.5 mm Well Width
- 13 min., 85 max. μL Sample Volume
Specifications
Specifications
| For Use With (Equipment) | SUB13 Midi Submarine Gel Electrophoresis Unit |
| Type | Comb |
| No. of Wells | 10 |
| Thickness (English) | 0.03 in. |
| Thickness (Metric) | 0.1 cm |
| Volume (Fill) Metric | 40 μL |
| Width (English) Well | 0.37 in. |
| Width (Metric) Well | 0.95 cm |
| Depth (English) Pan | 0.19 in. |
| Depth (Metric) Pan | 0.5 cm |