Learn More
Hoefer™ SUB20 Maxi-Standard and SUB20C Maxi-Cooled Submarine Combs
Accessories for SUB20 Maxi-Standard and SUB20C Maxi-Cooled Submarines
Brand:  Hoefer™ SUB20-C1.5-20MC
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12685766
  • 59.05€ / Each
Estimated Shipment
13-07-2021
Specifications
0.19 in.
SUB20 Maxi Standard and SUB20C Maxi Cooled Submarine
20
0.15 cm
0.27 in.
0.5 cm
Comb
0.05 in.
45 μL
0.7 cm